Assay Details
Target Gene Details
Entrez Gene ID: | 1024 |
Gene Name: | cyclin dependent kinase 8 |
Gene Aliases: |
K35 |
Location: |
Chr.13:26254104-26405238 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK8 | NM_001260.2 | NP_001251.1 | ||
| NM_001318368.1 | NP_001305297.1 | |||
| XM_011534864.1 | XP_011533166.1 |
More Information
Set Membership: |
|

Genomic Map