Assay Details
Target Gene Details
Entrez Gene ID: | 10910 |
Gene Name: | SGT1 homolog, MIS12 kinetochore complex assembly cochaperone |
Gene Aliases: |
SGT1 |
Location: |
Chr.13:52652691-52700756 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGT1 | NM_001130912.2 | NP_001124384.1 | ||
| NM_001320831.1 | NP_001307760.1 | |||
| NM_006704.4 | NP_006695.1 | |||
| XM_011534891.2 | XP_011533193.1 | |||
| XM_011534893.2 | XP_011533195.1 | |||
| XM_017020365.1 | XP_016875854.1 | |||
| XM_017020366.1 | XP_016875855.1 | |||
| XM_017020367.1 | XP_016875856.1 | |||
| XM_017020368.1 | XP_016875857.1 | |||
| AF132856.1 | AAD30062.1 | |||
| AJ344097.1 | CAC51433.1 | |||
| AK291402.1 | ||||
| AK292128.1 | ||||
| AK302363.1 | ||||
| AY271314.1 | AAQ01749.1 | |||
| AY321358.1 | AAQ76039.1 | |||
| BC000911.1 | AAH00911.1 | |||
| BT009798.1 | AAP88800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044949 | Chr.13:52497005 - 52715291 on Build GRCh38 | Gain |
|
| dgv1653n100 | Chr.13:52564997 - 52728939 on Build GRCh38 | Gain |
|
| nsv820041 | Chr.13:52487203 - 52706453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map