Assay Details
Target Gene Details
Entrez Gene ID: | 646982 |
Gene Name: | long intergenic non-protein coding RNA 598 |
Gene Aliases: |
TTL |
Location: |
Chr.13:40347132-40481006 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00598 | NR_024506.1 | 6 | 2601 | |
| NR_024507.2 | 7 | 2741 | ||
| AY116215.1 | 2 | 2008 | AAM97781.1 | |
| AY116216.1 | 3 | 2146 | AAM97782.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1633n100 | Chr.13:40217600 - 40371437 on Build GRCh38 | Gain |
|

Genomic Map