Assay Details
Target Gene Details
Entrez Gene ID: | 7750 |
Gene Name: | zinc finger MYM-type containing 2 |
Gene Aliases: |
FIM, MYM, RAMP, SCLL, ZNF198 |
Location: |
Chr.13:19957412-20091845 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832557 | Chr.13:19910435 - 20081915 on Build GRCh38 | Gain |
|
| nsv1054226 | Chr.13:19511073 - 19989089 on Build GRCh38 | Gain |
|
| nsv561215 | Chr.13:19871303 - 20076371 on Build GRCh38 | Gain |
|
| esv2761244 | Chr.13:19389134 - 20043200 on Build GRCh38 | Gain |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
| esv3892297 | Chr.13:19803308 - 20022425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map