Assay Details
Target Gene Details
Entrez Gene ID: | 81617 |
Gene Name: | calcium binding protein 39 like |
Gene Aliases: |
MO25-BETA, MO2L, bA103J18.3 |
Location: |
Chr.13:49308628-49444346 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAB39L | NM_001079670.2 | NP_001073138.1 | ||
| NM_001287337.1 | NP_001274266.1 | |||
| NM_001287338.1 | NP_001274267.1 | |||
| NM_001287339.1 | NP_001274268.1 | |||
| NM_030925.3 | NP_112187.2 | |||
| XM_011535254.2 | XP_011533556.1 | |||
| XM_011535255.2 | XP_011533557.1 | |||
| XM_011535256.2 | XP_011533558.1 | |||
| XM_017020785.1 | XP_016876274.1 | |||
| XM_017020786.1 | XP_016876275.1 | |||
| XM_017020787.1 | XP_016876276.1 | |||
| XM_017020788.1 | XP_016876277.1 | |||
| AK022639.1 | BAB14147.1 | |||
| AK309011.1 | ||||
| AY288977.2 | ||||
| BC010993.2 | AAH10993.2 | |||
| BX647518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832607 | Chr.13:49182322 - 49346758 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map