Assay Details
Target Gene Details
Entrez Gene ID: | 100874255 |
Gene Name: | RNASEH2B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:50882378-50910712 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH2B-AS1 | NR_046552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv951085 | Chr.13:50882765 - 50891164 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79414511] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map