Assay Details
Target Gene Details
Entrez Gene ID: | 83548 |
Gene Name: | component of oligomeric golgi complex 3 |
Gene Aliases: |
SEC34 |
Location: |
Chr.13:45464895-45536698 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG3 | NM_031431.3 | NP_113619.2 | ||
| XM_011535266.1 | XP_011533568.1 | |||
| XM_011535267.2 | XP_011533569.1 | |||
| XM_017020790.1 | XP_016876279.1 | |||
| AF332595.1 | AAK06848.1 | |||
| AF349676.1 | AAK66974.1 | |||
| AK026305.1 | ||||
| AK225466.1 | ||||
| AK225952.1 | ||||
| AK304248.1 | ||||
| AK314387.1 | ||||
| BC038953.1 | AAH38953.1 | |||
| BX648211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| nsv510583 | Chr.13:45484456 - 45586401 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map