Assay Details
Target Gene Details
Entrez Gene ID: | 23281 |
Gene Name: | microtubule associated tumor suppressor candidate 2 |
Gene Aliases: |
CAZIP, ICIS, KIAA0774, TIP150 |
Location: |
Chr.13:28820314-29505947 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTUS2 | XM_011535019.2 | XP_011533321.1 | ||
| XM_011535020.2 | XP_011533322.1 | |||
| XM_011535021.2 | XP_011533323.1 | |||
| XM_017020500.1 | XP_016875989.1 | |||
| XM_017020501.1 | XP_016875990.1 | |||
| XM_017020502.1 | XP_016875991.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3390267 | Chr.13:28837956 - 28837966 on Build GRCh38 | Insertion |
|
| esv3386629 | Chr.13:28837954 - 28837973 on Build GRCh38 | Insertion |
|
| esv3308811 | Chr.13:28837951 - 28837952 on Build GRCh38 | Mobile Element Insertion |
|
| nsv832571 | Chr.13:28675794 - 28845425 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map