Assay Details
Target Gene Details
Entrez Gene ID: | 51028 |
Gene Name: | vacuolar protein sorting 36 homolog |
Gene Aliases: |
C13orf9, CGI-145, EAP45 |
Location: |
Chr.13:52412602-52450678 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS36 | NM_001282168.1 | NP_001269097.1 | ||
| NM_001282169.1 | NP_001269098.1 | |||
| NM_016075.3 | NP_057159.2 | |||
| AF151903.1 | AAD34140.1 | |||
| AI221133.1 | ||||
| AK023182.1 | BAB14451.1 | |||
| AK289740.1 | ||||
| BC037279.1 | AAH37279.1 | |||
| BC050439.1 | AAH50439.1 | |||
| BG400860.1 | ||||
| BX647847.1 | ||||
| CB959894.1 | ||||
| CR933653.1 | CAI45953.1 | |||
| DA461148.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428288 | Chr.13:52154235 - 52493997 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74534001,rs75543299] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map