Assay Details
Target Gene Details
Entrez Gene ID: | 87769 |
Gene Name: | gamma-glutamylamine cyclotransferase |
Gene Aliases: |
A2LD1 |
Location: |
Chr.13:100530164-100588792 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGACT | NM_001195087.1 | NP_001182016.1 | ||
| XM_017020804.1 | XP_016876293.1 | |||
| AK095850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037414 | Chr.13:100521000 - 100622872 on Build GRCh38 | Loss |
|
| esv3582760 | Chr.13:100471360 - 100566346 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map