Assay Details
Target Gene Details
Entrez Gene ID: | 646982 |
Gene Name: | long intergenic non-protein coding RNA 598 |
Gene Aliases: |
TTL |
Location: |
Chr.13:40347132-40481006 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00598 | NR_024506.1 | |||
| NR_024507.2 | ||||
| AY116215.1 | AAM97781.1 | |||
| AY116216.1 | AAM97782.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73171148] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map