Assay Details
Target Gene Details
Entrez Gene ID: | 57690 |
Gene Name: | trinucleotide repeat containing 6C |
Gene Aliases: |
- |
Location: |
Chr.17:77974984-78108835 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNRC6C | XM_006721996.3 | 1 | 737 | XP_006722059.3 |
| XM_006721997.3 | 1 | 737 | XP_006722060.3 | |
| XM_006722000.3 | 1 | 737 | XP_006722063.3 | |
| XM_017024906.1 | 1 | 737 | XP_016880395.1 | |
| XM_017024907.1 | 1 | 737 | XP_016880396.1 | |
| XM_017024908.1 | 1 | 737 | XP_016880397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828112 | Chr.17:77974861 - 77978031 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map