Assay Details
Target Gene Details
Entrez Gene ID: | 101928418 |
Gene Name: | uncharacterized LOC101928418 |
Gene Aliases: |
- |
Location: |
Chr.17:12671862-12706135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928418 | NR_104605.1 |
Target Gene Details
Entrez Gene ID: | 93649 |
Gene Name: | myocardin |
Gene Aliases: |
MYCD |
Location: |
Chr.17:12665890-12768949 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOCD | NM_001146312.2 | NP_001139784.1 | ||
| NM_153604.3 | NP_705832.1 | |||
| XM_005256863.1 | XP_005256920.1 | |||
| XM_005256864.1 | XP_005256921.1 | |||
| XM_017025342.1 | XP_016880831.1 | |||
| AF532596.1 | AAN33040.1 | |||
| AK128608.1 | ||||
| AK292885.1 | ||||
| AY764180.1 | AAV33439.1 | |||
| BC126307.1 | ||||
| BC143391.1 | ||||
| DA650534.1 | ||||
| DB226467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34095 | Chr.17:12436261 - 12841981 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113943669] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map