Assay Details
Target Gene Details
Entrez Gene ID: | 1770 |
Gene Name: | dynein axonemal heavy chain 9 |
Gene Aliases: |
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20 |
Location: |
Chr.17:11598431-11970168 on Build GRCh38 |
Assay Gene Location: | Within Intron 72 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAH9 | NM_001372.3 | NP_001363.2 | ||
| NM_004662.2 | NP_004653.2 | |||
| XM_011523703.1 | XP_011522005.1 | |||
| XM_017024292.1 | XP_016879781.1 | |||
| XM_017024293.1 | XP_016879782.1 | |||
| AB002355.2 | BAA21573.2 | |||
| AB290158.1 | ||||
| AF257737.1 | AAF69004.1 | |||
| AJ404468.1 | CAB94756.1 | |||
| BC128261.1 | ||||
| BC128421.1 | ||||
| BC153883.1 | ||||
| X99947.1 | CAA68207.1 |
Target Gene Details
Entrez Gene ID: | 7566 |
Gene Name: | zinc finger protein 18 |
Gene Aliases: |
HDSG1, KOX11, ZKSCAN6, ZNF535, ZSCAN38, Zfp535 |
Location: |
Chr.17:11957433-11997510 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF18 | XM_011524002.2 | XP_011522304.1 | ||
| XM_017025006.1 | XP_016880495.1 | |||
| XM_017025007.1 | XP_016880496.1 | |||
| XM_017025008.1 | XP_016880497.1 | |||
| XM_017025009.1 | XP_016880498.1 | |||
| XM_017025010.1 | XP_016880499.1 | |||
| XM_017025011.1 | XP_016880500.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76638363] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map