Assay Details
Target Gene Details
Entrez Gene ID: | 10609 |
Gene Name: | prolyl 3-hydroxylase family member 4 (non-enzymatic) |
Gene Aliases: |
LEPREL4, NO55, NOL55, SC65 |
Location: |
Chr.17:41801952-41812617 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3H4 | NM_006455.2 | 5 | 1285 | NP_006446.1 |
| XM_006721640.3 | 5 | 1703 | XP_006721703.1 | |
| AJ250583.1 | 5 | 1086 | CAC16786.1 | |
| AK056085.1 | 6 | 1468 | ||
| AK222945.1 | 5 | 1030 | BAD96665.1 | |
| AK301301.1 | 6 | 1002 | ||
| BC001047.2 | 5 | 1038 | AAH01047.1 | |
| BC007942.2 | 5 | 1033 | AAH07942.1 | |
| BC011701.2 | 5 | 1017 | AAH11701.1 | |
| U47621.1 | 5 | 1012 | AAC51792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060502 | Chr.17:41767306 - 41821751 on Build GRCh38 | Gain |
|
| dgv3174n100 | Chr.17:41778258 - 41843183 on Build GRCh38 | Gain |
|
| nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map