Assay Details
Target Gene Details
Entrez Gene ID: | 23580 |
Gene Name: | CDC42 effector protein 4 |
Gene Aliases: |
BORG4, CEP4, KAIA1777 |
Location: |
Chr.17:73283624-73312004 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP4 | NM_012121.4 | NP_036253.2 | ||
| XM_005257182.2 | XP_005257239.1 | |||
| AB042237.1 | BAB17272.1 | |||
| AK027385.1 | ||||
| AK097835.1 | ||||
| AK312536.1 | ||||
| BC002774.1 | AAH02774.1 | |||
| BC010451.1 | AAH10451.1 | |||
| BE387060.1 | ||||
| BI560730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833534 | Chr.17:73155642 - 73349355 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73996143] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map