Assay Details
Target Gene Details
Entrez Gene ID: | 433 |
Gene Name: | asialoglycoprotein receptor 2 |
Gene Aliases: |
ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2 |
Location: |
Chr.17:7101322-7115522 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| esv3639860 | Chr.17:7112919 - 7124549 on Build GRCh38 | Gain |
|
| esv2665200 | Chr.17:7091816 - 7333062 on Build GRCh38 | Deletion |
|
| nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map