Assay Details
Target Gene Details
Entrez Gene ID: | 92335 |
Gene Name: | STE20-related kinase adaptor alpha |
Gene Aliases: |
LYK5, NY-BR-96, PMSE, STRAD, Stlk |
Location: |
Chr.17:63702832-63741970 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
More Information
Additional Information:
For this assay, SNP(s) [rs73992363] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map