Assay Details
Target Gene Details
Entrez Gene ID: | 10900 |
Gene Name: | RUN domain containing 3A |
Gene Aliases: |
RAP2IP, RPIP-8, RPIP8 |
Location: |
Chr.17:44308559-44318670 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNDC3A | NM_001144825.1 | NP_001138297.1 | ||
| NM_001144826.1 | NP_001138298.1 | |||
| NM_006695.4 | NP_006686.1 | |||
| XM_005256964.3 | XP_005257021.1 | |||
| XM_017024035.1 | XP_016879524.1 | |||
| XM_017024036.1 | XP_016879525.1 | |||
| XM_017024037.1 | XP_016879526.1 | |||
| XM_017024038.1 | XP_016879527.1 | |||
| XM_017024039.1 | XP_016879528.1 | |||
| XM_017024040.1 | XP_016879529.1 | |||
| AB209802.1 | BAD93039.1 | |||
| AF055026.1 | AAC09366.1 | |||
| AK123330.1 | 1 | 575 | ||
| AK226045.1 | ||||
| AK313669.1 | ||||
| AL133657.1 | CAB63771.1 | |||
| BC006194.2 | AAH06194.1 | |||
| BX537443.1 | CAD97685.1 | |||
| CR456748.1 | CAG33029.1 | |||
| DA299778.1 | ||||
| U93871.1 | AAB68767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510713 | Chr.17:44201179 - 44340387 on Build GRCh38 | Deletion |
|
| nsv2061 | Chr.17:44310998 - 44345051 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map