Assay Details
Target Gene Details
Entrez Gene ID: | 101926996 |
Gene Name: | RUNDC3A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:44299574-44308431 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNDC3A-AS1 | NR_110802.1 | |||
| AK055254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510713 | Chr.17:44201179 - 44340387 on Build GRCh38 | Deletion |
|

Genomic Map