Assay Details
Target Gene Details
Entrez Gene ID: | 124930 |
Gene Name: | ankyrin repeat domain 13B |
Gene Aliases: |
- |
Location: |
Chr.17:29589769-29614761 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13B | NM_152345.4 | NP_689558.4 | ||
| XM_005257920.4 | XP_005257977.1 | |||
| XM_011524319.2 | XP_011522621.1 | |||
| XM_011524320.2 | XP_011522622.1 | |||
| XM_011524321.2 | XP_011522623.1 | |||
| XM_017024174.1 | XP_016879663.1 | |||
| XM_017024175.1 | XP_016879664.1 | |||
| AK092673.1 | ||||
| BC062597.1 | AAH62597.1 | |||
| DA445860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833409 | Chr.17:29465646 - 29655807 on Build GRCh38 | Gain |
|
| nsv827941 | Chr.17:29597091 - 29607641 on Build GRCh38 | Gain |
|

Genomic Map