Assay Details
Target Gene Details
Entrez Gene ID: | 84268 |
Gene Name: | RPA interacting protein |
Gene Aliases: |
HRIP, RIP |
Location: |
Chr.17:5419185-5433020 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPAIN | NM_001033002.3 | NP_001028174.2 | ||
| NM_001160244.1 | NP_001153716.1 | |||
| NM_001160246.1 | NP_001153718.1 | |||
| NM_001160266.1 | NP_001153738.1 | |||
| NR_027682.1 | ||||
| NR_027683.1 | ||||
| NR_027684.1 | ||||
| NR_027685.1 | ||||
| AA745783.1 | ||||
| AK098491.1 | 2 | 806 | BAC05317.1 | |
| AK226044.1 | ||||
| AY680654.1 | AAT80872.1 | |||
| AY680655.1 | AAT80873.1 | |||
| AY680656.1 | AAT80874.1 | |||
| AY680657.1 | AAT80875.1 | |||
| AY680658.1 | AAT80876.1 | |||
| AY680659.1 | AAT80877.1 | |||
| AY680660.1 | AAT80878.1 | |||
| AY775314.1 | AAX14368.1 | |||
| AY775315.1 | AAX14369.1 | |||
| AY775316.1 | AAX14370.1 | |||
| AY775317.1 | AAX14371.1 | |||
| AY775318.1 | AAX14372.1 | |||
| AY775319.1 | AAX14373.1 | |||
| AY775320.1 | AAX14374.1 | |||
| AY775321.1 | AAX14375.1 | |||
| AY775323.1 | AAX14377.1 | |||
| BC046349.1 | AAH46349.1 | |||
| BC051849.2 | AAH51849.1 | |||
| BI091755.1 | ||||
| CK005117.1 | ||||
| N41653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117113844] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map