Assay Details
Target Gene Details
Entrez Gene ID: | 162282 |
Gene Name: | ankyrin repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:56110935-56517016 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKFN1 | XM_011524430.2 | XP_011522732.1 | ||
| XM_011524431.2 | XP_011522733.1 | |||
| XM_017024263.1 | XP_016879752.1 | |||
| XM_017024270.1 | XP_016879759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2100 | Chr.17:55415019 - 56251155 on Build GRCh38 | Deletion |
|
| nsv575691 | Chr.17:56015458 - 56213790 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80074243] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map