Assay Details
Target Gene Details
Entrez Gene ID: | 9135 |
Gene Name: | rabaptin, RAB GTPase binding effector protein 1 |
Gene Aliases: |
RAB5EP, RABPT5 |
Location: |
Chr.17:5282263-5386340 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABEP1 | NM_001083585.2 | NP_001077054.1 | ||
| NM_001291581.1 | NP_001278510.1 | |||
| NM_001291582.1 | NP_001278511.1 | |||
| NM_004703.5 | NP_004694.2 | |||
| XM_011524060.1 | XP_011522362.1 | |||
| AF098638.1 | AAC70781.1 | |||
| AK297535.1 | ||||
| AK310932.1 | ||||
| AK314183.1 | ||||
| BC032435.1 | ||||
| BC041700.1 | AAH41700.1 | |||
| BI666786.1 | ||||
| DB169027.1 | ||||
| X77723.1 | ||||
| X91141.1 | CAA62580.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| dgv3089n100 | Chr.17:5187013 - 5327634 on Build GRCh38 | Gain |
|
| nsv1955 | Chr.17:5288751 - 5333334 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map