Assay Details
Target Gene Details
Entrez Gene ID: | 147179 |
Gene Name: | WAS/WASL interacting protein family member 2 |
Gene Aliases: |
WICH, WIRE |
Location: |
Chr.17:40219304-40284136 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WIPF2 | NM_133264.4 | NP_573571.1 | ||
| XM_005257083.2 | XP_005257140.1 | |||
| XM_005257084.2 | XP_005257141.1 | |||
| XM_011524412.1 | XP_011522714.1 | |||
| XM_011524413.1 | XP_011522715.1 | |||
| AF370396.1 | AAQ15232.1 | |||
| AK055057.1 | ||||
| AK289578.1 | ||||
| AK289779.1 | ||||
| AK290301.1 | ||||
| AK311544.1 | ||||
| AK315836.1 | ||||
| AL834239.1 | CAH56360.1 | |||
| BC065551.1 | AAH65551.1 | |||
| CN404607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668303 | Chr.17:40222659 - 40224406 on Build GRCh38 | Deletion |
|
| esv3640553 | Chr.17:40222698 - 40224276 on Build GRCh38 | Loss |
|
| nsv525796 | Chr.17:40150112 - 40248785 on Build GRCh38 | Gain |
|
| nsv833444 | Chr.17:40086570 - 40281028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map