Assay Details
Target Gene Details
Entrez Gene ID: | 9331 |
Gene Name: | beta-1,4-galactosyltransferase 6 |
Gene Aliases: |
B4Gal-T6, beta4Gal-T6 |
Location: |
Chr.18:31622246-31724670 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT6 | NM_004775.3 | NP_004766.2 | ||
| XM_005258387.4 | XP_005258444.1 | |||
| XM_006722578.3 | XP_006722641.1 | |||
| XM_006722579.3 | XP_006722642.1 | |||
| XM_017026088.1 | XP_016881577.1 | |||
| XM_017026089.1 | XP_016881578.1 | |||
| XM_017026090.1 | XP_016881579.1 | |||
| XM_017026091.1 | XP_016881580.1 | |||
| XM_017026092.1 | XP_016881581.1 | |||
| AB024742.2 | BAA76273.2 | |||
| AB209194.1 | BAD92431.1 | |||
| AF038664.1 | AAC39737.1 | |||
| AF069054.1 | AAC21570.1 | |||
| AF097159.1 | AAD41695.1 | |||
| AK314860.1 | ||||
| BC069620.1 | AAH69620.1 | |||
| BC069642.1 | AAH69642.1 | |||
| BC074835.2 | AAH74835.1 | |||
| BC074884.2 | AAH74884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893086 | Chr.18:31623572 - 31852661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map