Assay Details
Target Gene Details
Entrez Gene ID: | 55628 |
Gene Name: | zinc finger protein 407 |
Gene Aliases: |
- |
Location: |
Chr.18:74597735-75065672 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF407 | XM_005266726.4 | XP_005266783.1 | ||
| XM_006722500.3 | XP_006722563.1 | |||
| XM_011526068.2 | XP_011524370.1 | |||
| XM_011526069.2 | XP_011524371.1 | |||
| XM_011526070.1 | XP_011524372.1 | |||
| XM_017025838.1 | XP_016881327.1 | |||
| AK056288.1 | BAB71139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057441 | Chr.18:74344906 - 74669985 on Build GRCh38 | Gain |
|
| nsv577656 | Chr.18:74534683 - 75005167 on Build GRCh38 | Gain |
|
| esv3643124 | Chr.18:74623637 - 74695240 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map