Assay Details
Target Gene Details
Entrez Gene ID: | 89777 |
Gene Name: | serpin family B member 12 |
Gene Aliases: |
YUKOPIN |
Location: |
Chr.18:63519164-63569329 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB12 | XM_005266778.3 | XP_005266835.2 | ||
| XM_011526246.1 | XP_011524548.1 | |||
| XM_011526248.1 | XP_011524550.1 | |||
| XM_017026067.1 | XP_016881556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74882879] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map