Assay Details
Target Gene Details
Entrez Gene ID: | 79822 |
Gene Name: | Rho GTPase activating protein 28 |
Gene Aliases: |
- |
Location: |
Chr.18:6729718-6915716 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP28 | XM_005258144.1 | XP_005258201.2 | ||
XM_005258145.1 | XP_005258202.2 | |||
XM_005258146.2 | XP_005258203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059678 | Chr.18:6155115 - 6838291 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
