Assay Details
Target Gene Details
Entrez Gene ID: | 79839 |
Gene Name: | coiled-coil domain containing 102B |
Gene Aliases: |
ACY1L, AN, C18orf14, HsT1731 |
Location: |
Chr.18:68715221-69214350 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC102B | NM_001093729.1 | NP_001087198.1 | ||
| XM_017025973.1 | XP_016881462.1 | |||
| XM_017025975.1 | XP_016881464.1 | |||
| XM_017025976.1 | XP_016881465.1 | |||
| XM_017025979.1 | XP_016881468.1 | |||
| XM_017025980.1 | XP_016881469.1 | |||
| AK091108.1 | ||||
| AK308114.1 | ||||
| DC382329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058755 | Chr.18:68695641 - 68843121 on Build GRCh38 | Gain |
|
| esv3893127 | Chr.18:68641143 - 68863126 on Build GRCh38 | Gain |
|
| dgv3390n100 | Chr.18:68019389 - 68866022 on Build GRCh38 | Gain |
|
| esv2758736 | Chr.18:68308501 - 68725725 on Build GRCh38 | Gain+Loss |
|
| nsv577329 | Chr.18:68025058 - 68867575 on Build GRCh38 | Gain |
|
| nsv577369 | Chr.18:68713228 - 68826170 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73454069] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map