Assay Details
Target Gene Details
Entrez Gene ID: | 6563 |
Gene Name: | solute carrier family 14 member 1 (Kidd blood group) |
Gene Aliases: |
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE |
Location: |
Chr.18:45724123-45752520 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC14A1 | NM_001128588.3 | NP_001122060.3 | ||
| NM_001146036.2 | NP_001139508.2 | |||
| NM_001146037.1 | NP_001139509.1 | |||
| NM_001308278.1 | NP_001295207.1 | |||
| NM_001308279.1 | NP_001295208.1 | |||
| NM_015865.6 | NP_056949.4 | |||
| XM_005258329.1 | XP_005258386.1 | |||
| XM_005258333.1 | XP_005258390.1 | |||
| XM_006722526.3 | XP_006722589.1 | |||
| XM_011526142.1 | XP_011524444.1 | |||
| XM_011526144.1 | XP_011524446.1 | |||
| AF328892.1 | ||||
| AK091064.1 | ||||
| AK123681.1 | ||||
| AK127452.1 | ||||
| AK289608.1 | ||||
| AK294129.1 | ||||
| AK295268.1 | ||||
| BC050539.1 | AAH50539.1 | |||
| DA156178.1 | ||||
| GQ502682.1 | ||||
| HQ709264.1 | ||||
| KC677694.1 | ||||
| L36121.1 | ||||
| U35735.1 | AAB00181.1 | |||
| Y19039.1 | CAB60834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063292 | Chr.18:45715129 - 45737040 on Build GRCh38 | Gain |
|
| nsv525268 | Chr.18:45168400 - 45894830 on Build GRCh38 | Gain |
|
| esv3893100 | Chr.18:45542509 - 45858191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map