Assay Details
Target Gene Details
Entrez Gene ID: | 7776 |
Gene Name: | zinc finger protein 236 |
Gene Aliases: |
ZNF236A, ZNF236B |
Location: |
Chr.18:76822550-76970732 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF236 | NM_001306089.1 | NP_001293018.1 | ||
NM_007345.3 | NP_031371.3 | |||
XM_005266762.3 | XP_005266819.1 | |||
XM_011526165.2 | XP_011524467.1 | |||
XM_011526166.2 | XP_011524468.1 | |||
XM_011526168.2 | XP_011524470.1 | |||
XM_011526169.2 | XP_011524471.1 | |||
AF085243.1 | AAD55328.1 | |||
AF085244.1 | AAD55329.1 | |||
BC140859.1 | ||||
BX648028.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1056690 | Chr.18:76794730 - 76903712 on Build GRCh38 | Gain |
![]() ![]() |
nsv1063632 | Chr.18:76317786 - 77170410 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
