Assay Details
Target Gene Details
Entrez Gene ID: | 374868 |
Gene Name: | ATPase phospholipid transporting 9B (putative) |
Gene Aliases: |
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1 |
Location: |
Chr.18:79069275-79378391 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577794 | Chr.18:78951366 - 79472709 on Build GRCh38 | Gain |
|
| esv3643288 | Chr.18:79087927 - 79251969 on Build GRCh38 | Loss |
|
| nsv1065902 | Chr.18:78538045 - 80257174 on Build GRCh38 | Gain |
|
| esv3643290 | Chr.18:79088460 - 79140822 on Build GRCh38 | Loss |
|
| esv3643283 | Chr.18:79003881 - 79112789 on Build GRCh38 | Loss |
|
| esv2717497 | Chr.18:78170182 - 79635788 on Build GRCh38 | Deletion |
|
| nsv1141945 | Chr.18:78170394 - 79635820 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139116798] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map