Assay Details
Target Gene Details
Entrez Gene ID: | 284257 |
Gene Name: | biorientation of chromosomes in cell division 1 like 2 |
Gene Aliases: |
BOD1P, FAM44C |
Location: |
Chr.18:57147062-57150408 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOD1L2 | NM_001257964.1 | 1 | 2577 | NP_001244893.1 |
More Information
Set Membership: |
|

Genomic Map