Assay Details
Target Gene Details
Entrez Gene ID: | 11201 |
Gene Name: | DNA polymerase iota |
Gene Aliases: |
RAD30B, RAD3OB |
Location: |
Chr.18:54269479-54321266 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLI | NM_007195.2 | NP_009126.2 | ||
| XM_005258188.3 | XP_005258245.1 | |||
| XM_005258192.4 | XP_005258249.1 | |||
| XM_006722378.2 | XP_006722441.1 | |||
| XM_011525796.2 | XP_011524098.1 | |||
| XM_011525797.1 | XP_011524099.1 | |||
| AF140501.1 | AAD50381.1 | |||
| AF245438.1 | AAF63383.1 | |||
| AK093688.1 | ||||
| AK301578.1 | ||||
| AL136670.1 | CAB66605.1 | |||
| BC032662.1 | AAH32662.1 | |||
| DB450447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583078 | Chr.18:54271815 - 54278124 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map