Assay Details
Target Gene Details
Entrez Gene ID: | 103910 |
Gene Name: | myosin light chain 12B |
Gene Aliases: |
MLC-B, MRLC2 |
Location: |
Chr.18:3262113-3278284 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYL12B | NM_001144944.1 | NP_001138416.1 | ||
| NM_001144945.1 | NP_001138417.1 | |||
| NM_033546.3 | NP_291024.1 | |||
| AK291726.1 | ||||
| AV714583.1 | ||||
| AY320408.1 | AAP73808.1 | |||
| BC004994.1 | AAH04994.1 | |||
| BG696903.1 | ||||
| BG703257.1 | ||||
| CN302451.1 | ||||
| D50372.1 | BAA23323.1 | |||
| D82058.1 | BAB88918.1 | |||
| U26162.1 | AAA67367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641586 | Chr.18:3206430 - 3282328 on Build GRCh38 | Gain |
|
| esv3641585 | Chr.18:3200019 - 3415247 on Build GRCh38 | Gain |
|
| nsv1062247 | Chr.18:2887767 - 3394669 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115541514] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map