Assay Details
Target Gene Details
Entrez Gene ID: | 4087 |
Gene Name: | SMAD family member 2 |
Gene Aliases: |
JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2 |
Location: |
Chr.18:47833095-47931193 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD2 | NM_001003652.3 | 11 | 9943 | NP_001003652.1 |
| NM_001135937.2 | 10 | 9853 | NP_001129409.1 | |
| NM_005901.5 | 11 | 9835 | NP_005892.1 | |
| XM_005258259.3 | 12 | 10076 | XP_005258316.1 | |
| XM_006722451.3 | 12 | 10088 | XP_006722514.1 | |
| XM_011525984.2 | 11 | 9432 | XP_011524286.1 | |
| XM_011525985.2 | 13 | 10135 | XP_011524287.1 | |
| XM_017025745.1 | 13 | 10174 | XP_016881234.1 | |
| XM_017025746.1 | 11 | 9986 | XP_016881235.1 | |
| XM_017025747.1 | 12 | 10061 | XP_016881236.1 | |
| XM_017025748.1 | 13 | 10147 | XP_016881237.1 | |
| XM_017025750.1 | 12 | 10036 | XP_016881239.1 | |
| AY134745.1 | 1 | 472 | AAN08619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3347n100 | Chr.18:47833641 - 47881034 on Build GRCh38 | Loss |
|
| nsv1057722 | Chr.18:47817594 - 47874989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map