Assay Details
Target Gene Details
Entrez Gene ID: | 7276 |
Gene Name: | transthyretin |
Gene Aliases: |
CTS, CTS1, HEL111, HsT2651, PALB, TBPA |
Location: |
Chr.18:31591767-31599024 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTR | NM_000371.3 | NP_000362.1 | ||
| AF162690.1 | AAD45014.1 | |||
| AK312051.1 | ||||
| BC005310.1 | AAH05310.1 | |||
| BC020791.1 | AAH20791.1 | |||
| BT007189.1 | AAP35853.1 | |||
| CR456908.1 | CAG33189.1 | |||
| D00096.1 | BAA00059.1 | |||
| DB498675.1 | ||||
| DQ839490.1 | ||||
| EU794670.1 | ||||
| GU727633.1 | ||||
| K02091.1 | AAA60011.1 | |||
| M10605.1 | AAA60012.1 | |||
| M11714.1 | AAA61181.1 | |||
| U19780.1 | AAA73473.1 | |||
| X59498.1 | CAA42087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576636 | Chr.18:31566859 - 31610211 on Build GRCh38 | Loss |
|
| nsv1066881 | Chr.18:31569789 - 31600877 on Build GRCh38 | Gain |
|
| nsv833615 | Chr.18:31428433 - 31613013 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115314057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map