Assay Details
Target Gene Details
Entrez Gene ID: | 84735 |
Gene Name: | carnosine dipeptidase 1 |
Gene Aliases: |
CN1, CPGL2, HsT2308 |
Location: |
Chr.18:74534457-74585025 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNDP1 | NM_032649.5 | NP_116038.4 | ||
| AJ417564.1 | CAD10388.1 | |||
| AK098369.1 | ||||
| AK289916.1 | ||||
| AK303708.1 | ||||
| AK308716.1 | ||||
| AK314572.1 | ||||
| AY358756.1 | AAQ89116.1 | |||
| BC110295.1 | AAI10296.1 | |||
| BC113512.1 | ||||
| BC117122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057441 | Chr.18:74344906 - 74669985 on Build GRCh38 | Gain |
|
| esv3643117 | Chr.18:74454714 - 74614537 on Build GRCh38 | Loss |
|
| nsv577656 | Chr.18:74534683 - 75005167 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75541886] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map