Assay Details
Target Gene Details
Entrez Gene ID: | 6014 |
Gene Name: | Ras like without CAAX 2 |
Gene Aliases: |
RIBA, RIN, ROC2 |
Location: |
Chr.18:42743218-43115692 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIT2 | NM_001272077.1 | NP_001259006.1 | ||
| NM_002930.3 | NP_002921.1 | |||
| AF493922.1 | AAM12636.1 | |||
| AK313823.1 | ||||
| AY563951.1 | AAS75332.1 | |||
| BC018060.1 | AAH18060.1 | |||
| U71204.1 | AAB42214.1 | |||
| U78164.1 | AAB64245.1 | |||
| U78166.1 | AAB64247.1 | |||
| Y07565.1 | CAA68850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833632 | Chr.18:42607714 - 42808093 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map