Assay Details
Target Gene Details
Entrez Gene ID: | 374868 |
Gene Name: | ATPase phospholipid transporting 9B (putative) |
Gene Aliases: |
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1 |
Location: |
Chr.18:79069275-79378391 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577794 | Chr.18:78951366 - 79472709 on Build GRCh38 | Gain |
|
| nsv509704 | Chr.18:79012881 - 79077238 on Build GRCh38 | Insertion |
|
| nsv1065902 | Chr.18:78538045 - 80257174 on Build GRCh38 | Gain |
|
| esv3643283 | Chr.18:79003881 - 79112789 on Build GRCh38 | Loss |
|
| esv2717497 | Chr.18:78170182 - 79635788 on Build GRCh38 | Deletion |
|
| nsv1141945 | Chr.18:78170394 - 79635820 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map