Assay Details
Target Gene Details
Entrez Gene ID: | 5932 |
Gene Name: | RB binding protein 8, endonuclease |
Gene Aliases: |
COM1, CTIP, JWDS, RIM, SAE2, SCKL2 |
Location: |
Chr.18:22914121-23026486 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP8 | XM_006722519.2 | XP_006722582.1 | ||
| XM_006722520.2 | XP_006722583.1 | |||
| XM_011526132.2 | XP_011524434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2227 | Chr.18:22900840 - 22927879 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map