Assay Details
Target Gene Details
Entrez Gene ID: | 23255 |
Gene Name: | microtubule crosslinking factor 1 |
Gene Aliases: |
CCDC165, KIAA0802, SOGA2 |
Location: |
Chr.18:8705576-8832777 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTCL1 | XM_005258098.3 | XP_005258155.1 | ||
| XM_005258099.4 | XP_005258156.1 | |||
| XM_011525640.2 | XP_011523942.1 | |||
| XM_011525641.2 | XP_011523943.1 | |||
| XM_017025671.1 | XP_016881160.1 | |||
| XM_017025674.1 | XP_016881163.1 | |||
| AB018345.2 | BAA34522.2 |
More Information
Set Membership: |
|

Genomic Map