Assay Details
Target Gene Details
Entrez Gene ID: | 10892 |
Gene Name: | MALT1 paracaspase |
Gene Aliases: |
IMD12, MLT, MLT1, PCASP1 |
Location: |
Chr.18:58671386-58753806 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MALT1 | NM_006785.3 | NP_006776.1 | ||
| NM_173844.2 | NP_776216.1 | |||
| XM_011525794.1 | XP_011524096.1 | |||
| AB026118.1 | BAA83099.1 | |||
| AF130356.2 | AAD38507.2 | |||
| AF316597.1 | AAG38589.1 | |||
| AK291386.1 | ||||
| BC030143.2 | AAH30143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577050 | Chr.18:58664431 - 58732147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map