Assay Details
Target Gene Details
Entrez Gene ID: | 100287082 |
Gene Name: | gastric cancer associated transcript 2 (non-protein coding) |
Gene Aliases: |
MTCL1-AS1, MTCL1AS1 |
Location: |
Chr.18:8695856-8707621 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GACAT2 | NR_120598.1 | |||
| BM717007.1 | ||||
| BQ026945.1 |

Genomic Map