Assay Details
Target Gene Details
Entrez Gene ID: | 80323 |
Gene Name: | coiled-coil domain containing 68 |
Gene Aliases: |
SE57-1 |
Location: |
Chr.18:54901509-54959539 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC68 | NM_001143829.1 | NP_001137301.1 | ||
| NM_025214.2 | NP_079490.1 | |||
| XM_006722552.3 | XP_006722615.1 | |||
| XM_011526198.2 | XP_011524500.1 | |||
| XM_011526199.2 | XP_011524501.1 | |||
| XM_011526200.2 | XP_011524502.1 | |||
| XM_011526201.2 | XP_011524503.1 | |||
| XM_011526202.1 | XP_011524504.1 | |||
| XM_011526203.2 | XP_011524505.1 | |||
| XM_017026011.1 | XP_016881500.1 | |||
| AF273051.1 | AAG34911.1 | |||
| AK058097.1 | ||||
| AK313793.1 | ||||
| BC029508.1 | AAH29508.1 |
More Information
Set Membership: |
|

Genomic Map