Assay Details
Target Gene Details
Entrez Gene ID: | 440730 |
Gene Name: | tripartite motif containing 67 |
Gene Aliases: |
TNL |
Location: |
Chr.1:231162928-231221568 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM67 | NM_001004342.3 | NP_001004342.3 | ||
| NM_001300889.1 | NP_001287818.1 | |||
| XM_011544192.2 | XP_011542494.1 | |||
| XM_017001323.1 | XP_016856812.1 | |||
| AK126782.1 | BAC86689.1 | |||
| AY253917.1 | AAP69949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589106 | Chr.1:231110289 - 231278336 on Build GRCh38 | Gain |
|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| esv7591 | Chr.1:231167136 - 231167831 on Build GRCh38 | Loss |
|
| esv1972788 | Chr.1:231167146 - 231167823 on Build GRCh38 | Deletion |
|
| esv2494270 | Chr.1:231166914 - 231168385 on Build GRCh38 | Deletion |
|
| nsv1011066 | Chr.1:230940432 - 231242850 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73114165,rs73114169] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map