Assay Details
Target Gene Details
Entrez Gene ID: | 3707 |
Gene Name: | inositol-trisphosphate 3-kinase B |
Gene Aliases: |
IP3-3KB, IP3K, IP3K-B, IP3KB, PIG37 |
Location: |
Chr.1:226631690-226739327 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPKB | NM_002221.3 | NP_002212.3 | ||
| AJ242780.1 | CAC40650.1 | |||
| AK289700.1 | ||||
| AK313801.1 | ||||
| AY513281.1 | AAT08034.1 | |||
| BQ434930.1 | ||||
| X57206.1 | CAA40491.1 | |||
| Y18024.2 | CAB65055.3 |
Target Gene Details
Entrez Gene ID: | 100506443 |
Gene Name: | ITPKB intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.1:226656080-226675068 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPKB-IT1 | NR_103784.1 | |||
| BI116225.1 | ||||
| BI906918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522446 | Chr.1:226597829 - 226736237 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map