Assay Details
Target Gene Details
Entrez Gene ID: | 2035 |
Gene Name: | erythrocyte membrane protein band 4.1 |
Gene Aliases: |
4.1R, EL1, HE |
Location: |
Chr.1:28887091-29120046 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121709 | Chr.1:28878840 - 28908858 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map