Assay Details
Target Gene Details
Entrez Gene ID: | 3752 |
Gene Name: | potassium voltage-gated channel subfamily D member 3 |
Gene Aliases: |
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22 |
Location: |
Chr.1:111770662-111989577 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCND3 | NM_004980.4 | NP_004971.2 | ||
| NM_172198.2 | NP_751948.1 | |||
| XM_005270851.4 | XP_005270908.1 | |||
| XM_006710629.3 | XP_006710692.1 | |||
| XM_006710630.3 | XP_006710693.1 | |||
| XM_006710631.3 | XP_006710694.1 | |||
| XM_011541427.2 | XP_011539729.1 | |||
| XM_017001244.1 | XP_016856733.1 | |||
| AF048712.1 | AAC05121.1 | |||
| AF048713.1 | AAC05122.1 | |||
| AF120491.1 | AAD38898.1 | |||
| AF187963.1 | AAF01044.1 | |||
| AF187964.1 | AAF01045.1 | |||
| AF205856.1 | AAF20924.1 | |||
| AF205857.1 | AAF20925.1 | |||
| BC113475.1 | ||||
| BC113477.1 | ||||
| BC143390.1 |
Target Gene Details
Entrez Gene ID: | 100874295 |
Gene Name: | KCND3 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.1:111853762-111856905 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCND3-IT1 | NR_046783.1 | |||
| BX096614.1 |
More Information
Set Membership: |
|

Genomic Map